missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesprin 2 Monoclonal Antibody (K20-478-5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518075
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Nesprin 2 Monoclonal antibody specifically detects Nesprin 2 in Human samples. It is validated for Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot.Specifications
Nesprin 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8WXH0 | |
SYNE2 | |
Residues 1-459 of human Nesprin-2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
K20-478-5 | |
Unconjugated | |
SYNE2 | |
6820443O06Rik; AW546258; D12Ertd777e; EDMD5; KASH domain-containing protein 2; KASH2; KIAA1011; MGC96690; mKIAA1011; Nesp2; Nesp2g; nesprin 2; nesprin-2; NUA; NUANCE; nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; polytrophin; Protein NUANCE; spectrin repeat containing nuclear envelope protein 2; spectrin repeat containing, nuclear envelope 2; synaptic nuclear envelope 2; synaptic nuclear envelope protein 2; synaptic nuclei expressed gene 2; SYNE2; Syne-2; TROPH | |
Mouse | |
Protein G | |
RUO | |
23224 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |