missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA515337
Code : LE
Additional Details : Weight : 0.01000kg
Description
NEK2 Polyclonal antibody specifically detects NEK2 in Human samples. It is validated for Western Blot.Specifications
NEK2 | |
Polyclonal | |
Unconjugated | |
NEK2 | |
AA617254; C77054; HSPK 21; HsPK21; NEK 2; NEK 2A; Nek2; Nek2 kinase; NEK2A; Never in mitosis A-related kinase 2; NIMA - related expressed kinase 2; NIMA (never in mitosis gene a)-related expressed kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NIMA related kinase 2; nimA-like protein kinase 1; NIMA-related expressed kinase 2; NIMA-related kinase 2; nimA-related protein kinase 2; NLK 1; NLK1; PPP1R111; protein phosphatase 1, regulatory subunit 111; RP67; Serine/threonine-protein kinase Nek2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
4751 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P51955 | |
NEK2 | |
KLH conjugated synthetic peptide between 350-382 amino acids from the central region of human NEK2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |