missing translation for 'onlineSavingsMsg'
Learn More

Necdin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA544555

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15526542

  • £403.00 / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: VALSNRMPMT GLLLMILSLI YVKGRGARES AVWNVLRILG LRPWKKHSTF Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%

Necdin Polyclonal antibody specifically detects Necdin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Necdin
Polyclonal
Unconjugated
NDN
AI528698; HsT16328; Ndn; NECD; Necdin; necdin homolog; necdin, MAGE family member; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Peg6; Prader-Willi syndrome chromosome region; PWCR
Rabbit
Affinity Chromatography
RUO
4692
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q99608
NDN
Synthetic peptide directed towards the middle region of human NDN
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers