missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS8 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519344
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and human based on sequence homology.
NDUFS8 Polyclonal antibody specifically detects NDUFS8 in Mouse, Rat samples. It is validated for Western Blot.Specifications
NDUFS8 | |
Polyclonal | |
Unconjugated | |
NDUFS8 | |
BC021616; CI-23k; CI23KD; CI-23kD; complex I 23kDa subunit; complex I-23kD; NADH dehydrogenase (ubiquinone) Fe-S protein 8; NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S8; NADH-ubiquinone oxidoreductase 23 kDa subunit; NDUFS8; TYKY; TYKY subunit | |
Goat | |
Antigen affinity chromatography | |
RUO | |
225887, 293652 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8K3J1 | |
NDUFS8 | |
Synthetic peptide sequence (EAEIAANIQAD) corresponding to the internal amino acids of NDUFS8 | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |