missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 Isotype Control Cocktail, FITC, PE, Invitrogen™

Isotype Control Cocktail

Brand:  Invitrogen PA533180

Product Code. 15344381

  • £457.00 / 50 tests
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

PA5-33180 is suitable for use as a negative control for the measurement of non-specific binding of mouse monoclonals of isotype IgG1 in human tissues in a dual labelling technique using FITC and R-Phycoerythrin fluorochromes. This antibody contains a monoclonal mixture of FITC-conjugated Mouse IgG1 RPE-conjugated Mouse IgG1. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG1
Cocktail
PE
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Antigen not available at this time
50 Tests
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1
Control, Flow Cytometry
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Mouse
purified
RUO
Not Applicable
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers