missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meteorin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547494
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Meteorin Polyclonal antibody specifically detects Meteorin in Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Meteorin | |
Polyclonal | |
Unconjugated | |
METRN | |
1810034B16Rik; C16orf23; c380A1.2; hypoxia/reoxygenation regulatory factor; hypoxia/reoxygenation regulatory factor in astrocytes; Hyrac; Meteorin; meteorin, glial cell differentiation regulator; METRN; RGD1306222; RJD6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
70083 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8C1Q4 | |
METRN | |
Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1 Gly22-Asp291 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |