Learn More
MCT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14124
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4124 detects MCT-1 in mouse samples. PA1-4124 has been successfully used in Western blot procedure. By Western blot this antibody detects a ∽20 kDa protein representing MCT-1 in transfected 3T3 cells. The PA1-4124 immunogen is a synthetic peptide corresponding to residues K(3) K F D E K E N V S N C(14) of the 20 kDa MCT-1.
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.Specifications
MCT1 | |
Polyclonal | |
Unconjugated | |
P53986 | |
SLC16A1 | |
Synthetic peptide corresponding to residues K(3) K F D E K E N V S N C(14) of the 20 kDa MCT-1. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Slc16a1 | |
Multiple copies in a T-cell maligcies, Maligt T cell amplified sequence 1MCT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20501 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |