missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517112
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Guanine nucleotide exchange factor (GEF) that modulates the Rho family of GTPases. Promotes the conversion of some member of the Rho family GTPase from the GDP-bound to the GTP-bound form. Isoform 1 exhibits no activity toward RHOA, RAC1 or CDC42. Isoform 2 exhibits decreased GEF activity toward CDC42. Isoform 3 exhibits a weak but significant activity toward RAC1 and CDC42. Isoform 4 exhibits significant activity toward RHOA and CDC42. The truncated DBL oncogene is active toward RHOA, RAC1 and CDC42.Specifications
MCF2 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P10911 | |
MCF2 | |
Synthetic peptide corresponding to the amino terminus of human MCF2/Dbl | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MCF2 | |
ARHGEF21; B230117G22Rik; DBL; DBL-transforming protein; MCF.2 cell line derived transforming sequence; mcf.2 transforming sequence; MCF2; Mcf-2; MCF2-transforming protein; Oncogene MCF2 (oncogene DBL); OTTHUMP00000024159; proto-oncogene DBL; Proto-oncogene MCF-2; RGD1566098 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109904, 317598, 4168 | |
-20°C | |
Liquid |