missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCCC2 Monoclonal Antibody (2B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522463
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: AYSPRFLYIW PNARISVMGG EQAANVLATI TKDQRAREGK QFSSADEAAL KEPIIKKFEE EGNPYYSSAR VWDDGIIDPA DTRLVLGLSF SAALNAPIEK TDFGIFRM
MCCC2 Monoclonal antibody specifically detects MCCC2 in Human samples. It is validated for ELISA, Immunohistochemistry (PFA fixed), Western Blot.Specifications
MCCC2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9HCC0 | |
Mccc2 | |
MCCC2 (NP_071415, 456 a.a. ∽ 563 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
2B3 | |
Unconjugated | |
Mccc2 | |
3-methylcrotonyl-CoA carboxylase 2; 3-methylcrotonyl-CoA carboxylase non-biotin-containing subunit; 3-methylcrotonyl-CoA:carbon dioxide ligase subunit beta; 4930552N12Rik; biotin carboxylase; MCCase subunit beta; MCCB; Mccc2; meth; methylcrotonoyl-CoA carboxylase 2; methylcrotonoyl-CoA carboxylase 2 (beta); methylcrotonoyl-CoA carboxylase beta chain, mitochondrial; methylcrotonoyl-Coenzyme A carboxylase 2 (beta); non-biotin containing subunit of 3-methylcrotonyl-CoA carboxylase; testicular secretory protein Li 29 | |
Mouse | |
Affinity chromatography | |
RUO | |
64087 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |