missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H6N1 HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA141237
Code : LD
Additional Details : Weight : 0.01000kg
Description
An H6N1 influenza virus was isolated from a green-winged teal during the H5N1 outbreak in Hong Kong Special Administrative Region (SAR) in 1997. This virus possesses similar genes encoding internal proteins as in the human H5N1 and H9N2 influenza viruses. In 1999, influenza viruses from quail infected two humans in Hong Kong. suggesting the potential for avian influenza viruses to cross the species barrier and infect humans without prior reassortment in an intermediate host, such as the pig. The common features shared by H5N1 and H9N2 influenza viruses isolated from humans are the genes encoding the proteins of the replicating complex, the matrix protein (M) gene, the nonstructural protein (NS) gene, N1 neuraminidase (NA), This virus essentially represents the reemergence of the H5N1 influenza viruses with a different hemagglutinin (HA).Specifications
Influenza A H6N1 HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H6N1 HA; H6N1 hemagglutinin; Influenza H6N1; influenza hemagglutinin | |
Synthetic peptide corresponding to residues 128-143 of avian flu HA protein. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Rabbit | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG |