missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533309
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Panda, Dog, Bat, Bovine, Rabbit, Horse, Pig (100%).
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.Specifications
HTR4 | |
Polyclonal | |
Unconjugated | |
HTR4 | |
5 hydroxytryptamine ( serotonin) receptor 4; 5 hydroxytryptamine (serotonin) receptor 4; 5-HT<4L>; 5HT4; 5-HT4; 5-HT-4; 5HT4 receptor; 5-HT4 receptor; 5-HT4 receptor (two splicing variants L and S); 5-HT4R; 5HTR4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; Htr4; Serotonin receptor 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100034053, 100338471, 3360, 397431, 489198 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q13639, Q29006 | |
HTR4 | |
Synthetic 15 amino acid peptide from 3rd cytoplasmic domain of human 5HT4 Receptor. | |
50 μg | |
Primary | |
Canine, Equine, Human, Non-human Primate, Porcine, Rabbit | |
Antibody | |
IgG |