missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR1F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533301
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Monkey, Panda, Bat (100%).
5-HT1F is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. 5-HT1F is a member of the GPCR family. Expression of 5-HT1F has been reported in various brain structures, including cortex, hippocampus, trigeminal ganglia, and cerebral blood vessels. Expression in peripheral tissues is limited to uterus, mesentery, and artery. No expression has been detected in heart, kidney, liver, pancreas, or spleen.Specifications
HTR1F | |
Polyclonal | |
Unconjugated | |
HTR1F | |
5-HT-1E-beta; 5ht1f; 5-HT1F; 5-HT-1F; 5HT1F Receptor; 5-HT1f receptor; 5HT6; 5-hydroxytryptamine (serotonin) receptor 1E beta; 5-hydroxytryptamine (serotonin) receptor 1F; 5-hydroxytryptamine (serotonin) receptor 1F, G protein-coupled; 5-hydroxytryptamine receptor 1F; Htr1eb; HTR1EL; HTR1F; MR77; Serotonin receptor 1F; serotonin receptor gene (similar to Mm Htr1eb) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P30939 | |
HTR1F | |
Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human 5HT1F Receptor. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |