missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538926
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multi-protein complexes, the transportation of nascent poly-peptide chains across cellular membranes and the regulation of protein folding. HSPA14 (heat shock 70kDa protein 14), also known as HSP70-4 or HSP70L1, is a 509 amino acid novel HSP protein derived from human dendritic cells. Belonging to the heat shock protein 70 family, HSPA14 is thought to promote dendritic cell maturation. It is also suggested that HSPA14 stimulates secretion of the proinflammatory cytokines interleukin 12p70 (IL-12p70), IL-1 beta, TNF alpha, and the chemokines IP-10, MIP-1 alpha, MIP-1 beta and normal T cell expressed and secreted (RANTES).Specifications
HSPA14 | |
Polyclonal | |
Unconjugated | |
HSPA14 | |
Heat shock 70 kDa protein 14, HSP70-like protein 1Heat shock protein HSP60, HSPA14, HSP60, HSP70L1, HSP7E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
307133, 50497, 51182 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q0VDF9, Q99M31, Q6AYB4 | |
HSPA14 | |
A synthetic peptide derived from the internal region of human HSP7E | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |