missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSFX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546685
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AGAAQPAASM VMFPHLPALH HHCPHSHRTS QYMPASDGPQ AYPDYADQST Sequence homology: Human: 100%
HSFX1 Polyclonal antibody specifically detects HSFX1 in Human samples. It is validated for Western Blot.Specifications
HSFX1 | |
Polyclonal | |
Unconjugated | |
HSFX1 | |
heat shock transcription factor family, X linked 1; heat shock transcription factor family, X-linked 1; heat shock transcription factor, X-linked; HSFX1; HSFX2; LW-1; ORC4L; testicular secretory protein Li 21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100506164 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UBD0 | |
HSFX1 | |
Synthetic peptide directed towards the C-terminal of human HSFX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |