missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511725
This item is not returnable.
View return policy
Description
Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
Specifications
HGF | |
Polyclonal | |
Unconjugated | |
Hgf | |
C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3082 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
P14210 | |
Hgf | |
KLH conjugated synthetic peptide between 521-554 amino acids from the C-terminal region of human HGF | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HGF Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction