missing translation for 'onlineSavingsMsg'
Learn More

Hepatitis B Virus Surface Monoclonal Antibody (S 26), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA17603

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11595302

  • £394.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MA1-7603 detects HBsAg from virus samples. MA1-7603 has been successfully used in ELISA procedures. The MA1-7603 immunogen is HBsAg purified from human sera pool. Store at 4oC short term; store at -80oC long term, avoiding freeze-thaw cycles.

Hepatitis B Virus Surface Monoclonal antibody specifically detects Hepatitis B Virus Surface in Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot.
Specifications

Specifications

Hepatitis B Virus Surface
Monoclonal
1 mg/mL
PBS with 0.01% sodium azide
Mouse
Protein A
RUO
Virus
Antibody
IgG1
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
S 26
Unconjugated
HBsAg; HBV; hepatitis b virus surface antigen; HepB
HBsAg purified from human sera pool.
100 μg
Primary
4°C short term, -80°C long term
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers