missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBL, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MLST8 protein.
Brand: Abnova H00064223-B01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: MNTSPGTVGSDPVILATAGYDHTVRFWQAHSGICTRTVQHQDSQVNALEVTPDRSMIAAAGYQHIRMYDLNSNNPNPIISYDGVNKNIASVGFHEDGRWMYTGGEDCTARIWDLRSRNLQCQRIFQVNAPINCVCLHPNQAELIVGDQSGAIHIWDLKTDHNEQLIPEPEVSITSAHIDPDASYMAAVNSTGNCYVWNLTGGIGDEVTQLIPKTKIPAHTRYALQCRFSPDSTLLATCSADQTCKIWRTSNFSLMTELSIKSGNPGESSRGWMWGCAFSGDSQYIVTASSDNLARLWCVETGEIKREYGGHQKAVVCLAFNDSVLGSpecifications
GBL | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MLST8 protein. | |
MLST8 | |
GBL/LST8/POP3/WAT1/GbetaL | |
Mouse | |
50 μL | |
Yes | |
64223 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_022372 | |
MLST8 | |
MLST8 (NP_071767, 1 a.a. ∼ 326 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |