missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1099
Code : PN
Additional Details : Weight : 0.01000kg
Description
PA1-099 specifically detects GATA1 and does not cross-react with GATA2 or GATA3.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.Specifications
GATA1 | |
Polyclonal | |
Unconjugated | |
Gata1 | |
eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
Rabbit | |
Protein A | |
RUO | |
14460, 2623 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P15976, P17679 | |
Gata1 | |
N-terminal truncated human recombinant protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |