Learn More
Galectin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532251
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, HL-60, HCT116, mouse colon, LGALS4 transfected 293T lysate. Predicted reactivity: Dog (80%), Pig (81%), Rabbit (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.Specifications
Galectin 4 | |
Polyclonal | |
Unconjugated | |
Lgals4 | |
Antigen NY-CO-27; GAL4; gal-4; galectin 4; galectin-4; L-36; L-36 lactose-binding protein; L36LBI; L36LBP; lactose-binding lectin 4; Lectin galactose binding soluble 4 (Galectin-4); lectin, galactose binding, soluble 4; Lectin, galactose binding, soluble 4 (Galectin-4); lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; lectin, galactoside-binding, soluble, 4 (galectin 4); Lgals4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16855, 3960 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P56470, Q8K419 | |
Lgals4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 140 of Human Galectin4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |