missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin 2 Monoclonal Antibody (747307), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524357
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Galectin 2 Monoclonal antibody specifically detects Galectin 2 in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
Galectin 2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9CQW5 | |
LGALS2 | |
E. coli-derived recombinant mouse Galectin-2 Ser2-Glu130 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry (Frozen), Western Blot | |
747307 | |
Unconjugated | |
LGALS2 | |
2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2 | |
Rat | |
Protein A/G | |
RUO | |
107753 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |