missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525649
Code : LD
Additional Details : Weight : 0.01000kg
Description
Endocrine hormone of the central and peripheral nervous systems that binds and activates the G protein-coupled receptors GALR1, GALR2, and GALR3. This small neuropeptide may regulate diverse physiologic functions including contraction of smooth muscle of the gastrointestinal and genitourinary tract, growth hormone and insulin release and adrenal secretion. [Uniprot]Specifications
GAL | |
Polyclonal | |
Unconjugated | |
Gal | |
ETL8; GAL; GAL1; Galanin; galanin and GMAP prepropeptide; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin/GMAP prepropeptide; galanin-message-associated peptide; galanin-related peptide; GAL-GMAP; GALN; GLNN; GMAP; MGC40167; preprogalanin | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
14419, 51083 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
P22466, P47212 | |
Gal | |
KLH conjugated synthetic peptide between 73-101 amino acids from the C-terminal region of human GAL | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |