Learn More
GABRB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540999
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TLDNRVADQL WVPDTYFLND KKSFVHGVTV KNRMIRLHPD GTVLYGLRIT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%
GABRB1 Polyclonal antibody specifically detects GABRB1 in Human samples. It is validated for Western Blot.Specifications
GABRB1 | |
Polyclonal | |
Unconjugated | |
Gabrb1 | |
AW061132; B230208N19Rik; GABA beta subunit; GABA(A) receptor; GABA(A) receptor subunit beta-1; GABA(A) receptor, beta 1; GABA(A)R beta-1; GABA-RB; Gabrb1; Gabrb-1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 1; gamma-aminobutyric acid (GABA) A receptor, subunit beta 1; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 1; gamma-aminobutyric acid A receptor beta 1; Gamma-aminobutyric acid receptor beta 1; gamma-aminobutyric acid receptor subunit beta-1; gamma-aminobutyric acid receptor, subunit beta 1; gamma-aminobutyric acid type A receptor beta1 subunit; gamma-aminobutyric acid type A receptor subunit beta1; GARB1; GBRB1; si:dkey-20i10.4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2560 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P18505 | |
Gabrb1 | |
Synthetic peptide directed towards the N-terminal of human GABRB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |