missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542252
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VDPNILCPLG SQEAPKPSGK EDLTNISPFP QPPQKDEGSN CSEDKVVESL Sequence homology: Human: 100%
FOXR2 Polyclonal antibody specifically detects FOXR2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FOXR2 | |
Polyclonal | |
Unconjugated | |
FOXR2 | |
EG436240; Forkhead box protein N6; forkhead box protein R2; forkhead box R2; Foxn6; Foxr2 | |
Rabbit | |
Protein A | |
RUO | |
139628 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6PJQ5 | |
FOXR2 | |
Synthetic peptide directed towards the N-terminal of human FOXR2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |