Learn More
FOXJ1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541090
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LGALEALELS PPLSPASHVD VDLTIHGRHI DCPATWGPSV EQAADSLDFD Sequence homology: Cow: 93%; Dog: 92%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 100%; Rat: 100%
FOXJ1 Polyclonal antibody specifically detects FOXJ1 in Human samples. It is validated for Western Blot.Specifications
FOXJ1 | |
Polyclonal | |
Unconjugated | |
FOXJ1 | |
FKHL13; FKHL-13; fork head homologue 4; forkhead box J1; forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; Foxj1; Hepatocyte nuclear factor 3 forkhead homolog 4; Hfh4; HFH-4; HNF-3/forkhead homolog 4; MGC35202 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2302 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92949 | |
FOXJ1 | |
Synthetic peptide directed towards the middle region of human FOXJ1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |