missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXTL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526058
Code : LD
Additional Details : Weight : 0.01000kg
Description
EXTL3 is probable glycosyltransferase.Specifications
EXTL3 | |
Polyclonal | |
Unconjugated | |
EXTL3 | |
2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2137 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
O43909 | |
EXTL3 | |
KLH conjugated synthetic peptide between 43-72 amino acids from the N-terminal region of human EXTL3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |