Learn More
EXTL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530025
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EXTL3 is probable glycosyltransferase.Specifications
EXTL3 | |
Polyclonal | |
Unconjugated | |
EXTL3 | |
2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43909 | |
EXTL3 | |
Recombinant fragment corresponding to a region within amino acids 667 and 915 of Human EXTL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |