Learn More
EXTL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530024
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (87%), Rat (85%), Pig (85%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.Specifications
EXTL2 | |
Polyclonal | |
Unconjugated | |
EXTL2 | |
3000001D04Rik; alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; AW146439; exostoses (multiple)-like 2; exostoses-like 2; exostosin like glycosyltransferase 2; exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXTL2; Extr2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9UBQ6 | |
EXTL2 | |
Recombinant fragment corresponding to a region within amino acids 81 and 300 of Human Exostosin-like 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |