Learn More
EXOSC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541861
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PLEIVFGMNG RIWVKAKTIQ QTLILANILE ACEHMTSDQR KQIFSRLAES Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 100%; Rat: 92%
EXOSC3 Polyclonal antibody specifically detects EXOSC3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
EXOSC3 | |
Polyclonal | |
Unconjugated | |
Exosc3 | |
2310005D06Rik; AI593501; bA3J10.7; CGI-102; Exosc3; Exosome complex component RRP40; exosome complex exonuclease RRP40; exosome complex exonuclease RRP40; exosome complex component RRP40; exosome component 3; hRrp-40; hRrp40p; hypothetical protein LOC533245; LOW QUALITY PROTEIN: exosome complex component RRP40; p10; PCH1B; Ribosomal RNA-processing protein 40; RP11-3J10.8; RRP40; Rrp40p | |
Rabbit | |
Protein A | |
RUO | |
51010 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NQT5 | |
Exosc3 | |
Synthetic peptide directed towards the C-terminal of human EXOSC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |