Learn More
Enterovirus 71 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532202
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Vero cell lysate infected with EV-71, EV71 C2 infected RD, EV71 B4 infected RD. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Entervirus 71 is a potentially fatal virus first isolated in 1969. Enterovirus 71 has been found in the spinal cord and medulla and can produce a range of symptoms from rashes to meningitis and ecephalitis.Specifications
Enterovirus 71 | |
Polyclonal | |
Unconjugated | |
EV71 | |
Enterovius 71 | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
5.6 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |