Learn More
ESRRA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528749
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, Mouse kidney, Rat kidney, A549. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Estrogen-related receptor alpha 1 (ERR alpha, ERR1) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. This protein binds to an ERR-alpha response element (ERRE) containing a single consensus half-site, 5'-tnaaggtca-3' can bind to the medium-chain acyl coenzyme a dehydrogenase (mcad) response element nrre-1 and may act as an important regulator of mcad promoter. Studies have shown that ERR1 functions a metabolic regulator and is often expressed in tissues that preferentially metabolize fatty acids. The corresponding gene for the ERR1 is NR3B1.Specifications
ESRRA | |
Polyclonal | |
Unconjugated | |
ESRRA | |
ERR alpha; ERR1; ERRa; ERRalpha; ERR-alpha; Errra; ESRL1; ESRRA; ESRRA1; ESRRA2; estrogen receptor related 1; estrogen receptor-like 1; estrogen related receptor alpha; estrogen related receptor alpha 1; estrogen related receptor, alpha; estrogen-related nuclear receptor alpha; estrogen-related receptor alpha; estrogen-related receptor, alpha; Estrra; HERR1; LOW QUALITY PROTEIN: steroid hormone receptor ERR1; NR3B1; Nuclear receptor subfamily 3 group B member 1; orphan nuclear receptor; steroid hormone receptor ERR1; Steroid hormone receptor ERR1 (Estrogen-related receptor, alpha) (ERR-alpha) (Estrogen receptor-like 1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2101, 26379, 293701 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.32 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O08580, P11474, Q5QJV7 | |
ESRRA | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human ERR alpha | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |