Learn More
ERH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521388
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, H1299, HCT116, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (99%), Xenopus laevis (100%), Pig (100%), Chicken (98%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May have a role in the cell cycle. [UniProt]Specifications
ERH | |
Polyclonal | |
Unconjugated | |
ERH | |
DROER; Enhancer of rudimentary homolog; enhancer of rudimentary homolog (Drosophila); Erh; Mer; Prei1; preimplantation protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13877, 2079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.79 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P84089, P84090 | |
ERH | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 86 (P84090) of ERH | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |