missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOPA Decarboxylase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518373
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
DOPA Decarboxylase (aromatic L-amino acid decarboxylase, AADC; DDC) catalyzes the second reaction in the biosynthesis of catecholamines and serotonin (Waymire and Haycock, 2002; Berry et al., 1996; Haycock et al., 2003). It is also involved in the biosynthesis of trace amines. DDC antibodies can therefore be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999).Specifications
DOPA Decarboxylase | |
Polyclonal | |
Unconjugated | |
DDC | |
AADC; aromatic L-amino acid decarboxylase; aromatic-L-amino acid decarboxylase; aromatic-L-amino-acid decarboxylase; DDC; DOPA decarboxylase; dopa decarboxylase (aromatic L-amino acid decarboxylase); HGNC:2719; L-Dopa decarboxylase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P20711 | |
DDC | |
Synthetic peptide sequence (WEHIKELAADVL) corresponding to the C-terminus amino acids of DDC | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |