missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-DLL4, APC, Clone: 447506, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | DLL4 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human DLL4, Ser27-Pro524, Accession # Q9NR61 |
Clone | 447506 |
Applications | Flow Cytometry |
Conjugate | APC |
Specifications
DLL4 | |
447506 | |
APC | |
Q9NR61 | |
IgG2a | |
54567 | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human DLL4, Ser27-Pro524, Accession # Q9NR61 | |
Flow Cytometry | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Rat | |
Primary | |
Detects human DLL4 in direct ELISAs. | |
Protein A or G purified from hybridoma culture supernatant |
For Research Use Only