Learn More
DHDDS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543253
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: NRRYAKKCQV ERQEGHSQGF NKLAETLRWC LNLGILEVTV YAFSIENFKR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%
DHDDS Polyclonal antibody specifically detects DHDDS in Human samples. It is validated for Western Blot.Specifications
DHDDS | |
Polyclonal | |
Unconjugated | |
DHDDS | |
3222401G21Rik; cis-IPTase; cis-isoprenyltransferase; cis-prenyl transferase; Cis-prenyltransferase subunit hCIT; CIT; CPT; dedol-PP synthase; dehydrodolichyl diphosphate syntase complex subunit Dhdds; dehydrodolichyl diphosphate synthase; dehydrodolichyl diphosphate synthase complex subunit DHDDS; dehydrodolichyl diphosphate synthase isoform a; dehydrodolichyl diphosphate synthase subunit; Dhdds; DS; epididymis tissue protein Li 189m; FLJ13102; HDS; RP59; W91638; wu:fd56c06; zgc:77088 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79947 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86SQ9 | |
DHDDS | |
Synthetic peptide directed towards the N-terminal of human DHDDS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |