Learn More
Dendritic Cells Rat anti-Mouse, Clone: MIDC-8, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA182248
Code : LD
Additional Details : Weight : 0.01000kg
Description
Rat anti Mouse dendritic/interdigitating cells antibody, clone MIDC-8 recognizes an antigen in the cytoplasm of rat dendritic cells as distinct spots near the nucleus. Rat anti Mouse dendritic/interdigitating cells antibody, clone MIDC-8 does not stain thymic epithelial cells which are the major contamits in thymic dendritic cell preparations.
In normal tissue this antibody identifies dendritic cells and a proportion of B lymphocytes. Evidence in pathological tissue and functional studies suggests it binds to an epitope expressed by antigen presenting cells. Within the macrophage/dendritic cell populations the epitope seen by RFD1 is coexpressed with RFD7 by a subset of cells that exhibit suppressive activity on T cell stimulation.Specifications
Dendritic Cells | |
Monoclonal | |
Unconjugated | |
Rat | |
2 mL | |
Primary | |
4°C or -20°C if preferred | |
Liquid |
Immunohistochemistry (Frozen) | |
MIDC-8 | |
DCs; DC | |
Proprietary Immunogen. | |
RUO | |
Mouse | |
Antibody | |
IgG2a |