Learn More
Cytosine Deaminase Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185365
Code : LD
Additional Details : Weight : 0.01000kg
Description
In Western blot, this antibody detects a band at ∽13 and 17 kDa. Sheep anti cytosine deaminase antibody recognizes cytosine deaminase, also known as cytosine aminohydrolase.
Cytosine Deaminase Polyclonal antibody specifically detects Cytosine Deaminase in Yeast samples. It is validated for ELISA, Western Blot.Specifications
Cytosine Deaminase | |
Polyclonal | |
Unconjugated | |
FCY1 | |
Cytosine aminohydrolase; Cytosine deaminase; FCY1; YP9499.17; YPR062W | |
Sheep | |
Protein G | |
RUO | |
856175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
5 mg/mL | |
PBS with 0.09% sodium azide | |
Q12178 | |
FCY1 | |
Purified yeast cytosine deaminase. | |
1 mL | |
Primary | |
Yeast | |
Antibody | |
IgG |