missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen II Monoclonal Antibody (2B1.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA137493
Code : LD
Additional Details : Weight : 1.24740kg
Description
Staining of formalin/parafin tissues requires disgestion of tissue sections with pepsin at 1 mg/mL Tris-HCl, pH 2.0 for 15 min at RT or 10 min at 37C.
COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.Specifications
Collagen II | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02458, P02459, P02460, P05539, P28481 | |
Col2a1 | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
200 μg | |
Primary | |
Bovine, Chicken, Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2B1.5 | |
Unconjugated | |
Col2a1 | |
alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha 1 type IIA collagen precursor; alpha collagen type II; alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CB11; CG2A1A; CGA; Chondrocalcin; COL11A3; Col2; Col2a; col2a1; Col2a-1; col2a1.L; col2a1a; col2a1-a; col2a1-b; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen type II alpha 1 chain; collagen type IIA; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); collagen, type II, alpha 1 L homeolog; COLLII; cyanogen bromide; Del1; disproportionate micromelia; Dmm; Lpk; M100856; MGC131516; pro alpha1 type II collagen; pro-alpha 1(II); pro-alpha-1 type II collagen; procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; Rgsc856; SEDC; STL1; type II collagen; type II collagen cyanogen bromide fra | |
Mouse | |
Protein A | |
RUO | |
1280, 12824, 25412, 395069, 407142 | |
4° C | |
Liquid |