missing translation for 'onlineSavingsMsg'
Learn More

CLA, Rat anti-Human, Clone: HECA-452, BV605, BD

Brand:  BD Biosciences 563960

Additional Details : Weight : 0.01000kg

Product Code. 15873239

  • £635.00 / Each
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Rat
Clone: HECA-452
Isotype: IgM, κ
Species Reactivity: Hu
Immunogen: Lymphocyte-depleted Stromal Preparation of Tonsil

Flow cytometry

Specifications

Specifications

CLA
HECA-452
Lymphocyte-depleted Stromal Preparation of Tonsil
100 Tests
Primary
Monoclonal
Rat
Affinity Purified
RUO
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers