Learn More
CD14 Mouse anti-Human, FITC, Clone: M5E2, BD
Mouse Monoclonal Antibody
£325.00 - £427.00
Specifications
Antigen | CD14 |
---|---|
Clone | M5E2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15828848
|
BD Biosciences
557153 |
50 Tests |
£325.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15808138
|
BD Biosciences
555397 |
100 Tests |
£427.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with the 53-55kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Host Species: Mouse
Clone: M5E2
Isotype: IgG2a, κ
Cross Reactivity: Human, Rhesus, Cynomolgus, Baboon, Dog
Flow Cytometry
Specifications
CD14 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
M5E2 | |
Monoclonal | |
Mouse | |
Immunology | |
Human CD14 Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.