missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf59 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546251
This item is not returnable.
View return policy
Description
Peptide sequence: EKAVRVRRGI TKAGERDKAP SLKSRSIVTS SGTTASAPPH SPGQAGGHAS Sequence homology: Human: 100%
C16orf59 Polyclonal antibody specifically detects C16orf59 in Human samples. It is validated for Western Blot.Specifications
C16orf59 | |
Polyclonal | |
Unconjugated | |
TEDC2 | |
C16orf59; chromosome 16 open reading frame 59; FLJ13909; TEDC2; Tubulin epsilon and delta complex protein 2; uncharacterized protein C16orf59 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
80178 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7L2K0 | |
TEDC2 | |
Synthetic peptide directed towards the N-terminal of human C16orf59 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |