Learn More
Annexin V Mouse anti-Human, CoraLite 647, Clone: 1E6A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-66245-100UL
Additional Details : Weight : 0.00010kg
Description
Annexin A5 (ANXA5), is a member of the structurally related family of annexin proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII.
Specifications
Annexin V | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunofluorescence, Flow Cytometry | |
1E6A8 | |
CoraLite 647 | |
100 μL | |
308 | |
Liquid |