Learn More
Ammonium thiocyanate, 99+%, extra pure, Thermo Scientific Chemicals
£35.38 - £419.09
Chemical Identifiers
CAS | 1762-95-4 |
---|---|
Molecular Formula | CH4N2S |
Molecular Weight (g/mol) | 76.12 |
MDL Number | MFCD00011428 |
InChI Key | SOIFLUNRINLCBN-UHFFFAOYSA-N |
Synonym | ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt |
PubChem CID | 15666 |
IUPAC Name | azanium;thiocyanate |
SMILES | C(#N)[S-].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144930
|
Thermo Scientific Acros
206505000 |
500 g | Plastic bottle |
£35.38
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553761
|
Thermo Scientific Acros
206500010 |
1 kg | Plastic bottle |
£61.81
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10414415
|
Thermo Scientific Acros
206500025 |
2.5 kg | Plastic bottle |
£131.66
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15189233
|
Thermo Scientific Acros
206500100 |
10 kg | Plastic Drum |
£419.09
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1762-95-4 | |
76.12 | |
SOIFLUNRINLCBN-UHFFFAOYSA-N | |
15666 | |
C(#N)[S-].[NH4+] |
CH4N2S | |
MFCD00011428 | |
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
azanium;thiocyanate |
Specifications
149.0°C | |
White | |
Crystals | |
CH4N2S | |
MFCD00011428 | |
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
SOIFLUNRINLCBN-UHFFFAOYSA-N | |
azanium;thiocyanate | |
15666 | |
≥99% | |
1.3 | |
Ammonium thiocyanate |
1762-95-4 | |
4.0 to 6.0 (5% aq. soln.) | |
99% min. (on dried basis) (Argentometry) | |
NH4SCN | |
15,558 | |
Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate | |
C(#N)[S-].[NH4+] | |
76.12 | |
76.12 | |
Extra Pure | |
1.3000g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-175-6
- Ammonium thiocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only