missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium magnesium phosphate hexahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
It is used as pharmaceutical intermediae.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediae.
Solubility
Insoluble in water.
Notes
Store in a cool place. Ensure proper ventilation.
It is used as pharmaceutical intermediae.
Solubility
Insoluble in water.
Notes
Store in a cool place. Ensure proper ventilation.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Formula Weight | 245.41 (137.32 Anhydrous) |
Percent Purity | 98% |
Density | 1.711 |
Chemical Name or Material | Ammonium magnesium phosphate hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'