missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium dihydrogen phosphate, 98%, Thermo Scientific Chemicals
22.90 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Use promo code "21742" to get your promotional price.
Click to view available options
Quantity:
500 g
2500 g
10000 g
Description
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.
Notes
Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.
Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.
Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.
Notes
Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 198°C to 204°C |
Quantity | 500 g |
Merck Index | 14,543 |
Solubility Information | Soluble in water. Slightly soluble in alcohol. Insoluble in acetone. |
IUPAC Name | azanium;dihydrogen phosphate |
Formula Weight | 115.03 |
Percent Purity | 98% |
Odor | Odorless |
Density | 1.803 |
Chemical Name or Material | Ammonium dihydrogen phosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'