missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate Solution According to European Pharmacopoeia (EP), Reagecon
High-purity, stable product developed and tested for European pharmacopoeia (EP) applications including the preparation of other reagents (4.1.1), identification reactions of ions and functional groups (2.3.1), and as a reagent in the performance of tests outlined in several EP monographs.
Brand: Reagecon 1005201
This item is not returnable.
View return policy
Description
- European Pharmacopoeia Reagent, Solution
- High-purity stable product developed and tested for European pharmacopoeia (EP) applications.
- Used for the preparation of other reagents (4.1.1), identification reactions of ions and functional groups (2.3.1) and as a reagent in the performance of tests outlined in several EP monographs
- This Reagecon EP reagent is accurate, stable and reproducible.
- Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
Amber Glass Winchester Bottle GL45 | |
1 L | |
EP Testing | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ammonium carbonate solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : ammonium carbonate