missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 10 Kda
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
£326.00
Specifications
Disposable | Single-use |
---|---|
MWCO | 10 kDa |
Sterility | Non-sterile |
Housing Material | Regenerated Cellulose (RC) |
Type | Centrifugal Filters |
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
15292220
|
Merck
ACS501024 |
24 Amicon Ultra NMWCO 10 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap | 24Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.Specifications
Single-use | |
Non-sterile | |
Centrifugal Filters |
10 kDa | |
Regenerated Cellulose (RC) | |
12 cm |