missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ TE 70 and TE 70 PWR Semi-Dry Transfer Unit
Electrotransfer proteins from polyacrylamide gels up to 14 x 16cm (TE 77 PWR) and 21 x 26cm (TE 77) in size in less than one hour
Brand: Cytiva TE70
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
- Requires minimal buffer, only enough to saturate blotting papers and membranes
- Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
- Suitable for gels up to 14 × 16 cm
- Tested for use with Amersham™ ECL Western Blotting reagents
- Automatic stopping feature; the transfer is stopped when buffer gets depleted, preventing overheating
- TE 70 does not come with a power supply
Specifications
ECL Semi-Dry Blotters are designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. | |
14 x 16 cm | |
TE 70 | |
9 cm |
TE 70 Semi-Dry Transfer Unit, 14 × 16 cm | |
25 sheets blotting paper, 50 sheets cellophane, 2 solid masks, masks for 7 x 8 cm and 14 x 16 cm gels. | |
38 cm |