missing translation for 'onlineSavingsMsg'
Learn More

Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

£31.10 - £495.00

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042259.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11369927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042259.18
50 g
£125.63
50g
Please to purchase this item. Need a web account? Register with us today!
11359927
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042259.30
250 g
£495.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629
[Al]

Specifications

7429-90-5
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
99.999%
Odorless
Slug
Aluminum slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Puratronic™
≈0.14g/slug
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only