Learn More
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
£43.22 - £239.19
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384658
|
Thermo Scientific Alfa Aesar
043022.18 |
50 g |
£43.22
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394658
|
Thermo Scientific Alfa Aesar
043022.30 |
250 g |
£82.61
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304668
|
Thermo Scientific Alfa Aesar
043022.A1 |
1 kg |
£239.19
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
[Al] |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 |
Specifications
Silver to Gray | |
Silver Gray powder | |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
-100+325 Mesh | |
Water reactive | |
Aluminum |
7429-90-5 | |
99.5% (Metals basis) | |
MFCD00134029 | |
400°C (752°F) | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Odorless | |
Powder |
Safety and Handling
P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
H250-H261-H500
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'rtecsNumber' : BD0330000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Aluminum powder
Signal Word
- Danger
Hazard Category
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P223-Do not allow contact with water.
- P231-Handle under inert gas.
- P232-Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P402+P404-Store in a dry place. Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Aluminium
RUO – Research Use Only