missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Potassium Sulfate Dodecahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Spezifikation
Quantity | 1 kg |
Linear Formula | AlK(SO4)2 · 12H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 474.39g/mol |
Percent Purity | ≥99.5% |
Grade | ACS/puriss. p.a. |
Density | 1.757g/mL at 25°C |
Chemical Name or Material | Aluminum Potassium Sulfate Dodecahydrate, Reag. Ph. Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'